missing translation for 'onlineSavingsMsg'
Learn More

1-Bromoundecane, 98%

CAS: 693-67-4 | C11H23Br | 235.209 g/mol

$60.55 - $633.74

Chemical Identifiers

CAS 693-67-4
Molecular Formula C11H23Br
Molecular Weight (g/mol) 235.209
MDL Number MFCD00000223
InChI Key IKPSIIAXIDAQLG-UHFFFAOYSA-N
Synonym undecyl bromide, undecane, 1-bromo, hendecyl bromide, n-undecyl bromide, bromoundecane, 1-bromo-undecane, n-undecyl-1-bromide, labotest-bb ltbb001165, undecylbromide, 1-bromanylundecane
PubChem CID 12744
IUPAC Name 1-bromoundecane
SMILES CCCCCCCCCCCBr
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1433314
View Documents
Thermo Scientific Chemicals
A1433314
25 g
Each for $60.55
Only null left
 
AAA1433322
View Documents
Thermo Scientific Chemicals
A1433322
100 g
Each for $180.05
Only null left
 
AAA1433336
View Documents
Thermo Scientific Chemicals
A1433336
500 g
Each for $633.74
Only null left
 
Description

Description

1-Bromoundecane is used in the preparation of Grignard reagent by reacting with Mg in THF (tetrahydrofuran). It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromoundecane is used in the preparation of Grignard reagent by reacting with Mg in THF (tetrahydrofuran). It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.
Specifications

Chemical Identifiers

693-67-4
235.209
IKPSIIAXIDAQLG-UHFFFAOYSA-N
12744
CCCCCCCCCCCBr
C11H23Br
MFCD00000223
undecyl bromide, undecane, 1-bromo, hendecyl bromide, n-undecyl bromide, bromoundecane, 1-bromo-undecane, n-undecyl-1-bromide, labotest-bb ltbb001165, undecylbromide, 1-bromanylundecane
1-bromoundecane

Specifications

693-67-4
1.052
>110°C (230°F)
1.4563
25 g
undecyl bromide, undecane, 1-bromo, hendecyl bromide, n-undecyl bromide, bromoundecane, 1-bromo-undecane, n-undecyl-1-bromide, labotest-bb ltbb001165, undecylbromide, 1-bromanylundecane
IKPSIIAXIDAQLG-UHFFFAOYSA-N
1-bromoundecane
12744
98%
-9°C
112°C to 114°C (10 mmHg)
C11H23Br
MFCD00000223
1098390
Insoluble in water.
CCCCCCCCCCCBr
235.209
235.22
1-Bromoundecane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H332-H335

EINECSNumber : 211-757-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only