missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromoundecane, 98%
Description
1-Bromoundecane is used in the preparation of Grignard reagent by reacting with Mg in THF (tetrahydrofuran). It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Bromoundecane is used in the preparation of Grignard reagent by reacting with Mg in THF (tetrahydrofuran). It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.
1-Bromoundecane is used in the preparation of Grignard reagent by reacting with Mg in THF (tetrahydrofuran). It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.
Specifications
Specifications
| Melting Point | -9°C |
| Density | 1.052 |
| Boiling Point | 112°C to 114°C (10 mmHg) |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.4563 |
| Quantity | 100 g |
| Beilstein | 1098390 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 235.22 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H332-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 211-757-3 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only