missing translation for 'onlineSavingsMsg'
Learn More

1-Bromotridecane 98.0+%, TCI America™

Catalog No. B09355ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B09355ML 5 mL
B093525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B09355ML Supplier TCI America Supplier No. B09355ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 765-09-3
Molecular Formula C13H27Br
Molecular Weight (g/mol) 263.26
MDL Number MFCD00000227
InChI Key BFDNZQUBFCYTIC-UHFFFAOYSA-N
Synonym Tridecyl Bromide
PubChem CID 13000
IUPAC Name 1-bromotridecane
SMILES CCCCCCCCCCCCCBr

Specifications

Color Yellow
Boiling Point 150°C
Quantity 5 mL
Formula Weight 263.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromotridecane
EINECSNumber (2)-0067
RTECSNumber YD3100000
TSCA Yes