Learn More
1-Bromotetradecane, 98%
CAS: 112-71-0 | C14H29Br | 277.29 g/mol
Supplier: Thermo Scientific Chemicals A160820E
Description
1-Bromotetradecane was used in the synthesis of tetradecylferrocene, cationic and zwitterionic gemini surfactants, and 9-tetradecylcarbazole. Also used for synthesis of metallomesogenic polymers based on bis[η1(N)-3,4-dialkyloxybenzylidene-4?-dodecyloxyaniline]dichloropalladium(II) with octyl, decyl, dodecyl, tetradecyl and hexadecyl alkyl groups.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromotetradecane was used in the synthesis of tetradecylferrocene, cationic and zwitterionic gemini surfactants, and 9-tetradecylcarbazole. Also used for synthesis of metallomesogenic polymers based on bis[η1(N)-3,4-dialkyloxybenzylidene-4′-dodecyloxyaniline]dichloropalladium(II) with octyl, decyl, dodecyl, tetradecyl and hexadecyl alkyl groups.
Solubility
Not miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
| 1-Bromotetradecane | |
| 4°C to 6°C | |
| 175°C to 178°C (20 mmHg) | |
| C14H29Br | |
| MFCD00000228 | |
| 1742640 | |
| Not miscible in water. | |
| CCCCCCCCCCCCCCBr | |
| 277.29 | |
| 277.3 |
| 112-71-0 | |
| 1.018 | |
| 158°C (316°F) | |
| 1.46 | |
| 2500 g | |
| myristyl bromide, tetradecyl bromide, tetradecane, 1-bromo, n-tetradecyl bromide, 1-tetradecyl bromide, bromotetradecane, n-tetradecyl-1-bromide, 1-bromo tetradecane, 1-bromo-n-tetradecane, tetradecane, bromo | |
| KOFZTCSTGIWCQG-UHFFFAOYSA-N | |
| 1-bromotetradecane | |
| 8208 | |
| 98% |
Chemical Identifiers
| 112-71-0 | |
| 277.29 | |
| KOFZTCSTGIWCQG-UHFFFAOYSA-N | |
| 8208 | |
| CCCCCCCCCCCCCCBr |
| C14H29Br | |
| MFCD00000228 | |
| myristyl bromide, tetradecyl bromide, tetradecane, 1-bromo, n-tetradecyl bromide, 1-tetradecyl bromide, bromotetradecane, n-tetradecyl-1-bromide, 1-bromo tetradecane, 1-bromo-n-tetradecane, tetradecane, bromo | |
| 1-bromotetradecane |
Safety and Handling
EINECSNumber : 203-999-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only