missing translation for 'onlineSavingsMsg'
Learn More

1-Bromotetradecane, 98%

Catalog No. p-7023039
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1608222 100 g
AAA1608236 500 g
AAA160820E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1608222 Supplier Thermo Scientific Chemicals Supplier No. A1608222

CAS: 112-71-0 | C14H29Br | 277.29 g/mol

1-Bromotetradecane was used in the synthesis of tetradecylferrocene, cationic and zwitterionic gemini surfactants, and 9-tetradecylcarbazole. Also used for synthesis of metallomesogenic polymers based on bis[η1(N)-3,4-dialkyloxybenzylidene-4?-dodecyloxyaniline]dichloropalladium(II) with octyl, decyl, dodecyl, tetradecyl and hexadecyl alkyl groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromotetradecane was used in the synthesis of tetradecylferrocene, cationic and zwitterionic gemini surfactants, and 9-tetradecylcarbazole. Also used for synthesis of metallomesogenic polymers based on bis[η1(N)-3,4-dialkyloxybenzylidene-4′-dodecyloxyaniline]dichloropalladium(II) with octyl, decyl, dodecyl, tetradecyl and hexadecyl alkyl groups.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 112-71-0
Molecular Formula C14H29Br
Molecular Weight (g/mol) 277.29
MDL Number MFCD00000228
InChI Key KOFZTCSTGIWCQG-UHFFFAOYSA-N
Synonym myristyl bromide, tetradecyl bromide, tetradecane, 1-bromo, n-tetradecyl bromide, 1-tetradecyl bromide, bromotetradecane, n-tetradecyl-1-bromide, 1-bromo tetradecane, 1-bromo-n-tetradecane, tetradecane, bromo
PubChem CID 8208
IUPAC Name 1-bromotetradecane
SMILES CCCCCCCCCCCCCCBr

Specifications

Melting Point 4°C to 6°C
Density 1.018
Boiling Point 175°C to 178°C (20 mmHg)
Flash Point 158°C (316°F)
Refractive Index 1.46
Quantity 100 g
Beilstein 1742640
Solubility Information Not miscible in water.
Formula Weight 277.3
Percent Purity 98%
Chemical Name or Material 1-Bromotetradecane
Show More Show Less
EINECSNumber 203-999-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only