missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromopentane 98.0+%, TCI America™
Supplier: TCI America B0628500ML
Specifications
| 1-Bromopentane | |
| Yellow | |
| C5H11Br | |
| 500 mL | |
| n-amyl bromide, pentyl bromide, amyl bromide, pentane, 1-bromo, n-pentyl bromide, 1-pentyl bromide, bromopentane, 1-bromo-pentane, 1-pentylbromide, unii-z2s4r599p0 | |
| CCCCCBr | |
| 151.047 | |
| 151.05 | |
| Liquid |
| 110-53-2 | |
| 130°C | |
| MFCD00000267 | |
| 1993 | |
| YZWKKMVJZFACSU-UHFFFAOYSA-N | |
| 1-bromopentane | |
| 8057 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 110-53-2 | |
| 151.047 | |
| YZWKKMVJZFACSU-UHFFFAOYSA-N | |
| 8057 | |
| CCCCCBr |
| C5H11Br | |
| MFCD00000267 | |
| n-amyl bromide, pentyl bromide, amyl bromide, pentane, 1-bromo, n-pentyl bromide, 1-pentyl bromide, bromopentane, 1-bromo-pentane, 1-pentylbromide, unii-z2s4r599p0 | |
| 1-bromopentane |
Safety and Handling
EINECSNumber : (2)-0067
RTECSNumber : RZ9770000
TSCA : Yes