missing translation for 'onlineSavingsMsg'
Learn More

1-Bromopentane 98.0+%, TCI America™

Catalog No. p-7118573
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B062825ML 25 mL
B0628500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B062825ML Supplier TCI America Supplier No. B062825ML
Only null left

Chemical Identifiers

CAS 110-53-2
Molecular Formula C5H11Br
Molecular Weight (g/mol) 151.047
MDL Number MFCD00000267
InChI Key YZWKKMVJZFACSU-UHFFFAOYSA-N
Synonym n-amyl bromide, pentyl bromide, amyl bromide, pentane, 1-bromo, n-pentyl bromide, 1-pentyl bromide, bromopentane, 1-bromo-pentane, 1-pentylbromide, unii-z2s4r599p0
PubChem CID 8057
IUPAC Name 1-bromopentane
SMILES CCCCCBr

Specifications

Color Yellow
Boiling Point 130°C
Quantity 25 mL
UN Number 1993
Formula Weight 151.05
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromopentane
EINECSNumber (2)-0067
RTECSNumber RZ9770000
TSCA Yes