missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromopentadecane 98.0+%, TCI America™
Supplier: TCI America B093625ML
Specifications
| 1-Bromopentadecane | |
| Yellow | |
| C15H31Br | |
| 25 mL | |
| JKOTZBXSNOGCIF-UHFFFAOYSA-N | |
| 1-bromopentadecane | |
| 12394 | |
| ≥98.0% (GC) |
| 629-72-1 | |
| 158°C | |
| MFCD00000229 | |
| pentadecyl bromide, pentadecane, 1-bromo, 1-bromo pentadecane, n-pentadecyl bromide, pentadecane,1-bromo, 1-bromopentadecane, acmc-209nb8, dsstox_cid_29296, dsstox_rid_83414, dsstox_gsid_49339 | |
| CCCCCCCCCCCCCCCBr | |
| 291.32 | |
| 291.32 | |
| Liquid |
Chemical Identifiers
| 629-72-1 | |
| 291.32 | |
| JKOTZBXSNOGCIF-UHFFFAOYSA-N | |
| 1-bromopentadecane |
| C15H31Br | |
| MFCD00000229 | |
| 12394 | |
| CCCCCCCCCCCCCCCBr |
Safety and Handling
EINECSNumber : (2)-0067
TSCA : Yes