accessibility menu, dialog, popup

UserName

H3CBr

CAS RN 629-72-1

IUPAC Name: 1-bromopentadecane
Synonyms: 1-bromopentadecane
Molecular Weight (g/mol): 291.32
Molecular Formula: C15H31Br
InChi Key: JKOTZBXSNOGCIF-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCBr
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)