missing translation for 'onlineSavingsMsg'
Learn More

1-Bromopentadecane, 97%

Catalog No. AAL0761809
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0761809 10 g
AAL0761818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0761809 Supplier Thermo Scientific Chemicals Supplier No. L0761809
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 629-72-1 | C15H31Br | 291.32 g/mol

1-Bromopentadecane has been used as regent in the synthesis of (S)-eicos-(4E)-en-1-yn-3-ol, a bioactive component of the marine sponge Cribrochalina vasculum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromopentadecane has been used as regent in the synthesis of (S)-eicos-(4E)-en-1-yn-3-ol, a bioactive component of the marine sponge Cribrochalina vasculum.

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible materials are oxidizing agents.

Chemical Identifiers

CAS 629-72-1
Molecular Formula C15H31Br
Molecular Weight (g/mol) 291.32
MDL Number MFCD00000229
InChI Key JKOTZBXSNOGCIF-UHFFFAOYSA-N
Synonym pentadecyl bromide, pentadecane, 1-bromo, 1-bromo pentadecane, n-pentadecyl bromide, pentadecane,1-bromo, 1-bromopentadecane, acmc-209nb8, dsstox_cid_29296, dsstox_rid_83414, dsstox_gsid_49339
PubChem CID 12394
IUPAC Name 1-bromopentadecane
SMILES CCCCCCCCCCCCCCCBr

Specifications

Melting Point 18°C to 19°C
Density 1.005
Boiling Point 159°C to 160°C (5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4608
Quantity 10 g
Beilstein 1744553
Solubility Information Not miscible in water.
Formula Weight 291.32
Percent Purity 97%
Chemical Name or Material 1-Bromopentadecane
Show More Show Less
EINECSNumber 211-104-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only