missing translation for 'onlineSavingsMsg'
Learn More

1-Bromohexadecane, 98%

CAS: 112-82-3 | C16H33Br | 305.34 g/mol

$44.85 - $257.74

Chemical Identifiers

CAS 112-82-3
Molecular Formula C16H33Br
Molecular Weight (g/mol) 305.34
MDL Number MFCD00000230
InChI Key HNTGIJLWHDPAFN-UHFFFAOYSA-N
Synonym hexadecyl bromide, cetyl bromide, n-hexadecyl bromide, hexadecane, 1-bromo, 1-hexadecyl bromide, n-hexadecyl-1-bromide, bromohexadecane, unii-76gji7gvam, hexadecane, bromo, 76gji7gvam
PubChem CID 8213
IUPAC Name 1-bromohexadecane
SMILES CCCCCCCCCCCCCCCCBr
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1514714
View Documents
Thermo Scientific Chemicals
A1514714
25 g
Each for $44.85
Only null left
 
AAA1514730
View Documents
Thermo Scientific Chemicals
A1514730
250 g
Each for $122.16
Only null left
 
AAA151470B
View Documents
Thermo Scientific Chemicals
A151470B
1000 g
Each for $257.74
Only null left
 
Description

Description

1-Bromohexadecane is used in the preparation of soluble carbon nano-onions by covalent functionalization with hexadecyl chains, as extraction solvent in determination of endocrine-disrupting phenols (EDPs) in water samples by ultrasound-assisted emulsification microextraction method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromohexadecane is used in the preparation of soluble carbon nano-onions by covalent functionalization with hexadecyl chains, as extraction solvent in determination of endocrine-disrupting phenols (EDPs) in water samples by ultrasound-assisted emulsification microextraction method.

Solubility
Difficult to mix.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications

Chemical Identifiers

112-82-3
305.34
HNTGIJLWHDPAFN-UHFFFAOYSA-N
8213
CCCCCCCCCCCCCCCCBr
C16H33Br
MFCD00000230
hexadecyl bromide, cetyl bromide, n-hexadecyl bromide, hexadecane, 1-bromo, 1-hexadecyl bromide, n-hexadecyl-1-bromide, bromohexadecane, unii-76gji7gvam, hexadecane, bromo, 76gji7gvam
1-bromohexadecane

Specifications

112-82-3
1
176°C (348°F)
C16H33Br
MFCD00000230
773989
Difficult to mix.
CCCCCCCCCCCCCCCCBr
305.34
305.35
1-Bromohexadecane
16°C to 18°C
336°C
Odorless
1.462
25 g
hexadecyl bromide, cetyl bromide, n-hexadecyl bromide, hexadecane, 1-bromo, 1-hexadecyl bromide, n-hexadecyl-1-bromide, bromohexadecane, unii-76gji7gvam, hexadecane, bromo, 76gji7gvam
HNTGIJLWHDPAFN-UHFFFAOYSA-N
1-bromohexadecane
8213
98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 204-008-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only