missing translation for 'onlineSavingsMsg'
Learn More

1-Bromohexadecane, 98%

CAS: 112-82-3 | C16H33Br | 305.34 g/mol

Supplier:  Thermo Scientific Chemicals A151470B

 View more versions of this product

Catalog No. AAA151470B


Only null left
Explore available promotions

Description

Description

1-Bromohexadecane is used in the preparation of soluble carbon nano-onions by covalent functionalization with hexadecyl chains, as extraction solvent in determination of endocrine-disrupting phenols (EDPs) in water samples by ultrasound-assisted emulsification microextraction method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromohexadecane is used in the preparation of soluble carbon nano-onions by covalent functionalization with hexadecyl chains, as extraction solvent in determination of endocrine-disrupting phenols (EDPs) in water samples by ultrasound-assisted emulsification microextraction method.

Solubility
Difficult to mix.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications

Specifications

1-Bromohexadecane
16°C to 18°C
336°C
Odorless
1.462
1000 g
hexadecyl bromide, cetyl bromide, n-hexadecyl bromide, hexadecane, 1-bromo, 1-hexadecyl bromide, n-hexadecyl-1-bromide, bromohexadecane, unii-76gji7gvam, hexadecane, bromo, 76gji7gvam
HNTGIJLWHDPAFN-UHFFFAOYSA-N
1-bromohexadecane
8213
98%
112-82-3
1
176°C (348°F)
C16H33Br
MFCD00000230
773989
Difficult to mix.
CCCCCCCCCCCCCCCCBr
305.34
305.35

Chemical Identifiers

112-82-3
305.34
HNTGIJLWHDPAFN-UHFFFAOYSA-N
8213
CCCCCCCCCCCCCCCCBr
C16H33Br
MFCD00000230
hexadecyl bromide, cetyl bromide, n-hexadecyl bromide, hexadecane, 1-bromo, 1-hexadecyl bromide, n-hexadecyl-1-bromide, bromohexadecane, unii-76gji7gvam, hexadecane, bromo, 76gji7gvam
1-bromohexadecane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 204-008-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only