missing translation for 'onlineSavingsMsg'
Learn More

1-Bromodecane 98.0+%, TCI America™

Supplier:  TCI America B0583500ML

 View more versions of this product

Catalog No. B0583500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1-Bromodecane
-30°C
139°C
MFCD00000221
decyl bromide, n-decyl bromide, decane, 1-bromo, 1-decyl bromide, decylbromide, bromodecane, decane, bromo, labotest-bb ltbb001164, decylmonobromide, n-decylbromide
CCCCCCCCCCBr
221.18
221.18
Liquid
112-29-8
Colorless
C10H21Br
500 mL
MYMSJFSOOQERIO-UHFFFAOYSA-N
1-bromodecane
8173
≥98.0% (GC)

Chemical Identifiers

112-29-8
221.18
MYMSJFSOOQERIO-UHFFFAOYSA-N
8173
CCCCCCCCCCBr
C10H21Br
MFCD00000221
decyl bromide, n-decyl bromide, decane, 1-bromo, 1-decyl bromide, decylbromide, bromodecane, decane, bromo, labotest-bb ltbb001164, decylmonobromide, n-decylbromide
1-bromodecane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0067

RTECSNumber : HD6850000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions