missing translation for 'onlineSavingsMsg'
Learn More

1-Bromodecane 98.0+%, TCI America™

Catalog No. p-7118541
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B058325ML 25 mL
B0583500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B058325ML Supplier TCI America Supplier No. B058325ML
Only null left

Chemical Identifiers

CAS 112-29-8
Molecular Formula C10H21Br
Molecular Weight (g/mol) 221.18
MDL Number MFCD00000221
InChI Key MYMSJFSOOQERIO-UHFFFAOYSA-N
Synonym decyl bromide, n-decyl bromide, decane, 1-bromo, 1-decyl bromide, decylbromide, bromodecane, decane, bromo, labotest-bb ltbb001164, decylmonobromide, n-decylbromide
PubChem CID 8173
IUPAC Name 1-bromodecane
SMILES CCCCCCCCCCBr

Specifications

Melting Point -30°C
Color Colorless
Boiling Point 139°C
Quantity 25 mL
Formula Weight 221.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromodecane
EINECSNumber (2)-0067
RTECSNumber HD6850000
TSCA Yes