missing translation for 'onlineSavingsMsg'
Learn More

1-Bromobutane 98.0+%, TCI America™

Catalog No. p-7118526
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B056025G 25 g
B0560500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B056025G Supplier TCI America Supplier No. B056025G
Only null left

Chemical Identifiers

CAS 109-65-9
Molecular Formula C4H9Br
Molecular Weight (g/mol) 137.02
MDL Number MFCD00000260
InChI Key MPPPKRYCTPRNTB-UHFFFAOYSA-N
Synonym butyl bromide, n-butyl bromide, bromobutane, butane, 1-bromo, 1-butyl bromide, butane, bromo, n-butylbromide, 1-bromo-butane, unii-sav6y78u3d, ccris 831
PubChem CID 8002
IUPAC Name 1-bromobutane
SMILES CCCCBr

Specifications

Melting Point -112°C
Color Colorless
Boiling Point 101°C
Quantity 25 g
UN Number 1126
Formula Weight 137.02
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromobutane
EINECSNumber (2)-0074
RTECSNumber EJ6225000
TSCA Yes