missing translation for 'onlineSavingsMsg'
Learn More

1-Bromobutane, 98%, Spectrum™ Chemical

Catalog No. p-7165153
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
2.5 kg
200 L
Packaging:
Amber Glass Bottle
Drum
Poly-Coated Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608081 25 g Amber Glass Bottle
18608082 250 g Amber Glass Bottle
18608080 200 L Drum
18608078 1 kg Amber Glass Bottle
18608079 2.5 kg Poly-Coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18608081 Supplier Spectrum Chemical Mfg Cor Supplier No. B115225GM
Only null left

Chemical Identifiers

CAS 109-65-9

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity 0.98
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.