missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromobutane, 98%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Packaging | Amber Glass Bottle |
| Quantity | 200 L |
| Molecular Weight (g/mol) | 137.03 |
| Percent Purity | 0.98 |
| Grade | Reagent |
| Physical Form | Liquid |
| Chemical Name or Material | 1-Bromobutane |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.