missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromoadamantane 99.0+%, TCI America™
$54.06 - $219.95
Chemical Identifiers
| CAS | 768-90-1 |
|---|---|
| Molecular Formula | C10H15Br |
| Molecular Weight (g/mol) | 215.134 |
| MDL Number | MFCD00074721 |
| InChI Key | VQHPRVYDKRESCL-UHFFFAOYSA-N |
| Synonym | 1-adamantyl bromide, adamantyl bromide, bromoadamantane, 1-bromo-adamantane, adamantane, 1-bromo, 1-bromotricyclo 3.3.1.13,7 decane, tricyclo 3.3.1.13,7 decane, 1-bromo, tricyclo 3.3.1.1 3,7-decane, 1-bromo, labotest-bb lt00239617, 3r,5s,7s-1-bromoadamantane |
| PubChem CID | 79106 |
| IUPAC Name | 1-bromoadamantane |
| SMILES | C1C2CC3CC1CC(C2)(C3)Br |
Chemical Identifiers
| 768-90-1 | |
| 215.134 | |
| VQHPRVYDKRESCL-UHFFFAOYSA-N | |
| 79106 | |
| C1C2CC3CC1CC(C2)(C3)Br |
| C10H15Br | |
| MFCD00074721 | |
| 1-adamantyl bromide, adamantyl bromide, bromoadamantane, 1-bromo-adamantane, adamantane, 1-bromo, 1-bromotricyclo 3.3.1.13,7 decane, tricyclo 3.3.1.13,7 decane, 1-bromo, tricyclo 3.3.1.1 3,7-decane, 1-bromo, labotest-bb lt00239617, 3r,5s,7s-1-bromoadamantane | |
| 1-bromoadamantane |
Specifications
| 768-90-1 | |
| White-Yellow | |
| MFCD00074721 | |
| 1-adamantyl bromide, adamantyl bromide, bromoadamantane, 1-bromo-adamantane, adamantane, 1-bromo, 1-bromotricyclo 3.3.1.13,7 decane, tricyclo 3.3.1.13,7 decane, 1-bromo, tricyclo 3.3.1.1 3,7-decane, 1-bromo, labotest-bb lt00239617, 3r,5s,7s-1-bromoadamantane | |
| C1C2CC3CC1CC(C2)(C3)Br | |
| 215.134 | |
| 215.13 | |
| Crystalline Powder |
| 119°C | |
| C10H15Br | |
| 25 g | |
| VQHPRVYDKRESCL-UHFFFAOYSA-N | |
| 1-bromoadamantane | |
| 79106 | |
| ≥99.0% (GC) | |
| 1-Bromoadamantane |
Safety and Handling
TSCA : No