missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromoadamantane, 99%
Description
1-Bromoadamantane is an important intermediate, used in synthesizing medicines and new materials etc.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Bromoadamantane is an important intermediate, used in synthesizing medicines and new materials etc.
Solubility
It is soluble in organic solvents and insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Moisture sensitive. Keep away from Strong oxidizing agents.
1-Bromoadamantane is an important intermediate, used in synthesizing medicines and new materials etc.
Solubility
It is soluble in organic solvents and insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Moisture sensitive. Keep away from Strong oxidizing agents.
Specifications
Specifications
| Melting Point | 117°C to 121°C |
| Quantity | 25 g |
| Beilstein | 1098857 |
| Solubility Information | It is soluble in organic solvents and insoluble in water. |
| Formula Weight | 215.14 |
| Percent Purity | 99% |
| Chemical Name or Material | 1-Bromoadamantane |
Safety and Handling
| EINECSNumber | 212-200-7 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only