missing translation for 'onlineSavingsMsg'
Learn More

1-Bromoadamantane, 99%

Catalog No. AAA1489314
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1489314 25 g
AAA1489322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1489314 Supplier Thermo Scientific Chemicals Supplier No. A1489314

CAS: 768-90-1 | C10H15Br | 215.134 g/mol

1-Bromoadamantane is an important intermediate, used in synthesizing medicines and new materials etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromoadamantane is an important intermediate, used in synthesizing medicines and new materials etc.

Solubility
It is soluble in organic solvents and insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Moisture sensitive. Keep away from Strong oxidizing agents.

Chemical Identifiers

CAS 768-90-1
Molecular Formula C10H15Br
Molecular Weight (g/mol) 215.134
MDL Number MFCD00074721
InChI Key VQHPRVYDKRESCL-UHFFFAOYSA-N
Synonym 1-adamantyl bromide, adamantyl bromide, bromoadamantane, 1-bromo-adamantane, adamantane, 1-bromo, 1-bromotricyclo 3.3.1.13,7 decane, tricyclo 3.3.1.13,7 decane, 1-bromo, tricyclo 3.3.1.1 3,7-decane, 1-bromo, labotest-bb lt00239617, 3r,5s,7s-1-bromoadamantane
PubChem CID 79106
IUPAC Name 1-bromoadamantane
SMILES C1C2CC3CC1CC(C2)(C3)Br

Specifications

Melting Point 117°C to 121°C
Quantity 25 g
Beilstein 1098857
Solubility Information It is soluble in organic solvents and insoluble in water.
Formula Weight 215.14
Percent Purity 99%
Chemical Name or Material 1-Bromoadamantane
EINECSNumber 212-200-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only