missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-8-iodonaphthalene 98.0+%, TCI America™
$259.11 - $892.89
Chemical Identifiers
| CAS | 4044-58-0 |
|---|---|
| Molecular Formula | C10H6BrI |
| Molecular Weight (g/mol) | 332.97 |
| MDL Number | MFCD11656276 |
| InChI Key | XNDLRPFISURYTA-UHFFFAOYSA-N |
| PubChem CID | 12347526 |
| IUPAC Name | 1-bromo-8-iodonaphthalene |
| SMILES | BrC1=C2C(I)=CC=CC2=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B5220200MG
|
TCI America
B5220200MG |
200 mg |
Each for $259.11
|
|
|||||
|
B52201G
|
TCI America
B52201G |
1 g |
Each for $892.89
|
|
|||||
Chemical Identifiers
| 4044-58-0 | |
| 332.97 | |
| XNDLRPFISURYTA-UHFFFAOYSA-N | |
| 1-bromo-8-iodonaphthalene |
| C10H6BrI | |
| MFCD11656276 | |
| 12347526 | |
| BrC1=C2C(I)=CC=CC2=CC=C1 |
Specifications
| 4044-58-0 | |
| 168°C | |
| MFCD11656276 | |
| XNDLRPFISURYTA-UHFFFAOYSA-N | |
| 1-bromo-8-iodonaphthalene | |
| 12347526 | |
| ≥98.0% (GC) | |
| 1-Bromo-8-iodonaphthalene |
| 101°C | |
| C10H6BrI | |
| 200 mg | |
| BrC1=C2C(I)=CC=CC2=CC=C1 | |
| 332.97 | |
| 332.97 | |
| Crystalline Powder |
Safety and Handling
TSCA : No