missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-8-iodonaphthalene 98.0+%, TCI America™

Catalog No. b5220200mg
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5220200MG 200 mg
B52201G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5220200MG Supplier TCI America Supplier No. B5220200MG

Chemical Identifiers

CAS 4044-58-0
Molecular Formula C10H6BrI
Molecular Weight (g/mol) 332.97
MDL Number MFCD11656276
InChI Key XNDLRPFISURYTA-UHFFFAOYSA-N
PubChem CID 12347526
IUPAC Name 1-bromo-8-iodonaphthalene
SMILES BrC1=C2C(I)=CC=CC2=CC=C1

Specifications

Melting Point 101°C
Boiling Point 168°C
Quantity 200 mg
Formula Weight 332.97
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromo-8-iodonaphthalene