accessibility menu, dialog, popup

UserName

BrI

CAS RN 4044-58-0

IUPAC Name: 1-bromo-8-iodonaphthalene
Synonyms: 1-bromo-8-iodonaphthalene
Molecular Weight (g/mol): 332.97
Molecular Formula: C10H6BrI
InChi Key: XNDLRPFISURYTA-UHFFFAOYSA-N
SMILES: BrC1=C2C(I)=CC=CC2=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results