missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-6-phenylhexane, 97%

Catalog No. AAL1456803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1456803 1 g
AAL1456806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1456803 Supplier Thermo Scientific Chemicals Supplier No. L1456803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 27976-27-8 | C12H17Br | 241.172 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27976-27-8
Molecular Formula C12H17Br
Molecular Weight (g/mol) 241.172
MDL Number MFCD00236022
InChI Key RAOLIGFNQJMMKW-UHFFFAOYSA-N
Synonym 6-bromohexyl benzene, 1-bromo-6-phenylhexane, benzene, 6-bromohexyl, 1-bromo-6-phenyl-hexane, 6-phenylhexyl bromide, hexane, 1-bromo-6-phenyl, 6-bromanylhexylbenzene, 5-benzylpentyl bromide, 6-bromo-1-phenylhexane
PubChem CID 561788
IUPAC Name 6-bromohexylbenzene
SMILES C1=CC=C(C=C1)CCCCCCBr

Specifications

Boiling Point 115°C to 118°C (6 mmHg)
Refractive Index 1.5295
Quantity 1 g
Beilstein 2207649
Formula Weight 241.18
Percent Purity 97%
Chemical Name or Material 1-Bromo-6-phenylhexane
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only