accessibility menu, dialog, popup

UserName

CAS RN 27976-27-8

Br

CAS RN 27976-27-8

IUPAC Name: (6-bromohexyl)benzene
Synonyms: (6-bromohexyl)benzene
Molecular Weight (g/mol): 241.17
Molecular Formula: C12H17Br
InChi Key: RAOLIGFNQJMMKW-UHFFFAOYSA-N
SMILES: BrCCCCCCC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)