missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-tert-butoxybenzene, 98%, Thermo Scientific Chemicals

Catalog No. p-7048049
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2015806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL2015806 Supplier Thermo Scientific Chemicals Supplier No. L2015806

It is used in the stereoselective synthesis of four stereoisomers of β-Methoxytyrosine, a component of callipeltin A.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the stereoselective synthesis of four stereoisomers of β-Methoxytyrosine, a component of callipeltin A.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 60876-70-2
Molecular Formula C10H13BrO
Molecular Weight (g/mol) 229.117
MDL Number MFCD00792676
InChI Key QIWQHUCUWNGYDZ-UHFFFAOYSA-N
Synonym 1-bromo-4-tert-butoxy benzene, 1-bromo-4-tert-butoxybenzene, 4-tert-butoxybromobenzene, 4-bromophenyl tert-butyl ether, 1-bromo-4-tert-butoxy-benzene, benzene, 1-bromo-4-1,1-dimethylethoxy, tert-butyl-4-bromophenyl ether, 1-bromo-4-2-methylpropan-2-yl oxy benzene, 1-tert-butoxy-4-bromobenzene, bromobutoxybenzene
PubChem CID 2763959
IUPAC Name 1-bromo-4-[(2-methylpropan-2-yl)oxy]benzene
SMILES CC(C)(C)OC1=CC=C(C=C1)Br

Specifications

Boiling Point 63°C to 65°C (0.4 mmHg)
Refractive Index 1.527
Quantity 5 g
Beilstein 2516411
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 229.12
Percent Purity 98%
Chemical Name or Material 1-Bromo-4-tert-butoxybenzene
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only