accessibility menu, dialog, popup

UserName

CAS RN 60876-70-2

IUPAC Name: 1-bromo-4-(tert-butoxy)benzene
Synonyms: 1-bromo-4-(tert-butoxy)benzene
Molecular Weight (g/mol): 229.12
Molecular Formula: C10H13BrO
InChi Key: QIWQHUCUWNGYDZ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC1=CC=C(Br)C=C1
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results