missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-propoxybenzene 98.0+%, TCI America™

Catalog No. p-7123066
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
B36395G 5 g
B363925G 25 g
2 options

Catalog No. B36395G

Supplier: TCI America B36395G

Only null left

Chemical Identifiers

CAS 39969-56-7
Molecular Formula C9H11BrO
Molecular Weight (g/mol) 215.09
MDL Number MFCD00156160
InChI Key VVPARGBRVKRZJC-UHFFFAOYSA-N
Synonym 4-n-propoxybromobenzene, 1-bromo-4-propoxylbenzene, 1-bromo-4-n-propoxybenzene, 1-bromo-4-n-propyloxybenzene, p-bromphenyl-n-propylather, acmc-209j9h, 1-bromo-4-propoxy-benzene, 1-4-bromophenoxy propane, 4-bromophenyl propyl ether, 1-bromanyl-4-propoxy-benzene
PubChem CID 2734198
IUPAC Name 1-bromo-4-propoxybenzene
SMILES CCCOC1=CC=C(C=C1)Br

Specifications

Color Yellow
Boiling Point 240°C
Quantity 5 g
Formula Weight 215.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-propoxybenzene
EINECSNumber (3)-0845
TSCA No