accessibility menu, dialog, popup

UserName

H3COBr

CAS RN 39969-56-7

IUPAC Name: 1-bromo-4-propoxybenzene
Synonyms: 1-bromo-4-propoxybenzene
Molecular Weight (g/mol): 215.09
Molecular Formula: C9H11BrO
InChi Key: VVPARGBRVKRZJC-UHFFFAOYSA-N
SMILES: CCCOC1=CC=C(Br)C=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)