missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-(phenylethynyl)benzene 98.0+%, TCI America™

Catalog No. B40241G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40241G 1 g
B40245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40241G Supplier TCI America Supplier No. B40241G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13667-12-4
Molecular Formula C14H9Br
Molecular Weight (g/mol) 257.13
MDL Number MFCD01596910
InChI Key XLHCHVUFUPJPEO-UHFFFAOYSA-N
Synonym 1-(4-Bromophenyl)-2-phenylacetylene
PubChem CID 290888
IUPAC Name 1-bromo-4-(2-phenylethynyl)benzene
SMILES C1=CC=C(C=C1)C#CC2=CC=C(C=C2)Br

Specifications

Melting Point 85°C
Color White-Yellow
Quantity 1 g
Formula Weight 257.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromo-4-(phenylethynyl)benzene