accessibility menu, dialog, popup

UserName

CAS RN 13667-12-4

IUPAC Name: 1-bromo-4-(2-phenylethynyl)benzene
Synonyms: 1-bromo-4-(2-phenylethynyl)benzene
Molecular Weight (g/mol): 257.13
Molecular Formula: C14H9Br
InChi Key: XLHCHVUFUPJPEO-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C#CC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results