missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-pentylbenzene 90.0+%, TCI America™

Catalog No. p-7119150
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16055G 5 g
B160525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16055G Supplier TCI America Supplier No. B16055G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 51554-95-1
Molecular Formula C11H15Br
Molecular Weight (g/mol) 227.145
MDL Number MFCD00061113
InChI Key SGCJPYYTVBHQGE-UHFFFAOYSA-N
Synonym 4-pentylbromobenzene, 4-bromo-n-pentylbenzene, 1-bromo-4-n-pentylbenzene, 4-n-amylbromobenzene, 4-n-pentylbromobenzene, 1-bromo-4-pentyl-benzene, p-bromopentylbenzene, 4-bromopentylbenzene, 1-amyl-4-bromobenzene, benzene, 1-bromo-4-pentyl
PubChem CID 2735599
IUPAC Name 1-bromo-4-pentylbenzene
SMILES CCCCCC1=CC=C(C=C1)Br

Specifications

Color Yellow
Boiling Point 148°C
Quantity 5 g
Formula Weight 227.15
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-pentylbenzene