accessibility menu, dialog, popup

UserName

CAS RN 51554-95-1

IUPAC Name: 1-bromo-4-pentylbenzene
Synonyms: 1-bromo-4-pentylbenzene
Molecular Weight (g/mol): 227.15
Molecular Formula: C11H15Br
InChi Key: SGCJPYYTVBHQGE-UHFFFAOYSA-N
SMILES: CCCCCC1=CC=C(Br)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)