missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-n-propoxybenzene, 98%

Catalog No. p-7081469
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1146206 5 g
AAA1146214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1146206 Supplier Thermo Scientific Chemicals Supplier No. A1146206

CAS: 39969-56-7 | C9H11BrO | 215.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39969-56-7
Molecular Formula C9H11BrO
Molecular Weight (g/mol) 215.09
MDL Number MFCD00156160
InChI Key VVPARGBRVKRZJC-UHFFFAOYSA-N
Synonym 4-n-propoxybromobenzene, 1-bromo-4-propoxylbenzene, 1-bromo-4-n-propoxybenzene, 1-bromo-4-n-propyloxybenzene, p-bromphenyl-n-propylather, acmc-209j9h, 1-bromo-4-propoxy-benzene, 1-4-bromophenoxy propane, 4-bromophenyl propyl ether, 1-bromanyl-4-propoxy-benzene
PubChem CID 2734198
IUPAC Name 1-bromo-4-propoxybenzene
SMILES CCCOC1=CC=C(C=C1)Br

Specifications

Density 1.356
Boiling Point 238°C to 240°C
Flash Point >110°C (230°F)
Quantity 5 g
Formula Weight 215.1
Percent Purity 98%
Chemical Name or Material 1-Bromo-4-n-propoxybenzene
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only