missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-n-pentylbenzene, 98%

Catalog No. AAA1445703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1445703 1 g
AAA1445706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1445703 Supplier Thermo Scientific Chemicals Supplier No. A1445703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 51554-95-1 | C11H15Br | 227.145 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51554-95-1
Molecular Formula C11H15Br
Molecular Weight (g/mol) 227.145
MDL Number MFCD00061113
InChI Key SGCJPYYTVBHQGE-UHFFFAOYSA-N
Synonym 4-pentylbromobenzene, 4-bromo-n-pentylbenzene, 1-bromo-4-n-pentylbenzene, 4-n-amylbromobenzene, 4-n-pentylbromobenzene, 1-bromo-4-pentyl-benzene, p-bromopentylbenzene, 4-bromopentylbenzene, 1-amyl-4-bromobenzene, benzene, 1-bromo-4-pentyl
PubChem CID 2735599
IUPAC Name 1-bromo-4-pentylbenzene
SMILES CCCCCC1=CC=C(C=C1)Br

Specifications

Quantity 1 g
Formula Weight 227.15
Percent Purity 98%
Chemical Name or Material 1-Bromo-4-n-pentylbenzene
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only