missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-ethylbenzene 98.0+%, TCI America™

Catalog No. B187925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B187925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B187925G Supplier TCI America Supplier No. B187925G

Chemical Identifiers

CAS 1585-07-5
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.064
MDL Number MFCD00000111
InChI Key URFPRAHGGBYNPW-UHFFFAOYSA-N
Synonym p-bromoethylbenzene, benzene, 1-bromo-4-ethyl, 4-ethylbromobenzene, p-ethylbromobenzene, 1-bromo-4-ethyl-benzene, unii-42li8tat6j, 1-ethyl-4-bromobenzene, 4-bromo-1-ethylbenzene, 4-ethyl-1-bromobenzene, 42li8tat6j
PubChem CID 15313
IUPAC Name 1-bromo-4-ethylbenzene
SMILES CCC1=CC=C(C=C1)Br

Specifications

Color Yellow
Boiling Point 204°C
Quantity 25 g
Formula Weight 185.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-ethylbenzene