accessibility menu, dialog, popup

UserName

CAS RN 1585-07-5

IUPAC Name: 1-bromo-4-ethylbenzene
Synonyms: 1-bromo-4-ethylbenzene
Molecular Weight (g/mol): 185.06
Molecular Formula: C8H9Br
InChi Key: URFPRAHGGBYNPW-UHFFFAOYSA-N
SMILES: CCC1=CC=C(Br)C=C1
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)