missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-ethylbenzene, 99%

Catalog No. p-7020902
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1301309 10 g
AAA1301318 50 g
AAA1301330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1301309 Supplier Thermo Scientific Chemicals Supplier No. A1301309

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1585-07-5 | C8H9Br | 185.064 g/mol

1-Bromo-4-ethylbenzene is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-4-ethylbenzene is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Avoid excessive heat and light.

Chemical Identifiers

CAS 1585-07-5
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.064
MDL Number MFCD00000111
InChI Key URFPRAHGGBYNPW-UHFFFAOYSA-N
Synonym p-bromoethylbenzene, benzene, 1-bromo-4-ethyl, 4-ethylbromobenzene, p-ethylbromobenzene, 1-bromo-4-ethyl-benzene, unii-42li8tat6j, 1-ethyl-4-bromobenzene, 4-bromo-1-ethylbenzene, 4-ethyl-1-bromobenzene, 42li8tat6j
PubChem CID 15313
IUPAC Name 1-bromo-4-ethylbenzene
SMILES CCC1=CC=C(C=C1)Br

Specifications

Density 1.341
Boiling Point 203°C to 204°C
Flash Point 63°C (145°F)
Refractive Index 1.544
Quantity 10 g
Beilstein 1856989
Solubility Information Insoluble in water.
Formula Weight 185.06
Percent Purity 99%
Chemical Name or Material 1-Bromo-4-ethylbenzene
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 216-439-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only