missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-4-dodecylbenzene 90.0+%, TCI America™
$169.41 - $508.25
Chemical Identifiers
| CAS | 126930-72-1 |
|---|---|
| Molecular Formula | C18H29Br |
| Molecular Weight (g/mol) | 325.334 |
| MDL Number | MFCD00191386 |
| InChI Key | RFFMRBISTZHCNY-UHFFFAOYSA-N |
| Synonym | 4-dodecylbromobenzene, 1-bromo-4-n-dodecylbenzene, benzene, 1-bromo-4-dodecyl, 4-bromododecylbenzene, pubchem22735, acmc-209bby, 1-bromo-4-dodecyl-benzene, ksc494g6d, 4-n-dodecylbromobenzene |
| PubChem CID | 15120506 |
| IUPAC Name | 1-bromo-4-dodecylbenzene |
| SMILES | CCCCCCCCCCCCC1=CC=C(C=C1)Br |
Chemical Identifiers
| 126930-72-1 | |
| 325.334 | |
| RFFMRBISTZHCNY-UHFFFAOYSA-N | |
| 15120506 | |
| CCCCCCCCCCCCC1=CC=C(C=C1)Br |
| C18H29Br | |
| MFCD00191386 | |
| 4-dodecylbromobenzene, 1-bromo-4-n-dodecylbenzene, benzene, 1-bromo-4-dodecyl, 4-bromododecylbenzene, pubchem22735, acmc-209bby, 1-bromo-4-dodecyl-benzene, ksc494g6d, 4-n-dodecylbromobenzene | |
| 1-bromo-4-dodecylbenzene |
Specifications
| 126930-72-1 | |
| 191°C | |
| MFCD00191386 | |
| 4-dodecylbromobenzene, 1-bromo-4-n-dodecylbenzene, benzene, 1-bromo-4-dodecyl, 4-bromododecylbenzene, pubchem22735, acmc-209bby, 1-bromo-4-dodecyl-benzene, ksc494g6d, 4-n-dodecylbromobenzene | |
| CCCCCCCCCCCCC1=CC=C(C=C1)Br | |
| 325.334 | |
| 325.33 | |
| Liquid |
| Yellow | |
| C18H29Br | |
| 5 g | |
| RFFMRBISTZHCNY-UHFFFAOYSA-N | |
| 1-bromo-4-dodecylbenzene | |
| 15120506 | |
| ≥90.0% (GC) | |
| 1-Bromo-4-dodecylbenzene |
Safety and Handling
TSCA : No