missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-4-cyclohexylbenzene 97.0+%, TCI America™
Supplier: TCI America B268625G
Specifications
| 1-Bromo-4-cyclohexylbenzene | |
| Yellow | |
| 25 g | |
| LVIJLEREXMVRAN-UHFFFAOYSA-N | |
| 1-bromo-4-cyclohexylbenzene | |
| 90718 | |
| ≥97.0% (GC) |
| 25109-28-8 | |
| C12H15Br | |
| p-bromocyclohexylbenzene, 1-brom-4-cyclohexylbenzol, zlchem 88, 4-cyclohexylbromobenzene, 4-bromophenylcyclohexane, acmc-1ccsm, 4-bromo-1-cyclohexylbenzene, 1-bromo-4-cyclohexyl-benzene, benzene, 1-bromo-4-cyclohexyl, lvijlerexmvran-uhfffaoysa | |
| C1CCC(CC1)C2=CC=C(C=C2)Br | |
| 239.156 | |
| 239.16 | |
| Liquid |
Chemical Identifiers
| 25109-28-8 | |
| 239.156 | |
| p-bromocyclohexylbenzene, 1-brom-4-cyclohexylbenzol, zlchem 88, 4-cyclohexylbromobenzene, 4-bromophenylcyclohexane, acmc-1ccsm, 4-bromo-1-cyclohexylbenzene, 1-bromo-4-cyclohexyl-benzene, benzene, 1-bromo-4-cyclohexyl, lvijlerexmvran-uhfffaoysa | |
| 1-bromo-4-cyclohexylbenzene |
| C12H15Br | |
| LVIJLEREXMVRAN-UHFFFAOYSA-N | |
| 90718 | |
| C1CCC(CC1)C2=CC=C(C=C2)Br |
Safety and Handling
TSCA : No