missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-cyclohexylbenzene, 98%

Catalog No. p-7044106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0463606 5 g
AAL0463614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0463606 Supplier Thermo Scientific Chemicals Supplier No. L0463606
Only null left

CAS: 25109-28-8 | C12H15Br | 239.156 g/mol

1-Bromo-4-cyclohexylbenzene is used as a organic light-emitting diode and as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-4-cyclohexylbenzene is used as a organic light-emitting diode and as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 25109-28-8
Molecular Formula C12H15Br
Molecular Weight (g/mol) 239.156
MDL Number MFCD00019339
InChI Key LVIJLEREXMVRAN-UHFFFAOYSA-N
Synonym p-bromocyclohexylbenzene, 1-brom-4-cyclohexylbenzol, zlchem 88, 4-cyclohexylbromobenzene, 4-bromophenylcyclohexane, acmc-1ccsm, 4-bromo-1-cyclohexylbenzene, 1-bromo-4-cyclohexyl-benzene, benzene, 1-bromo-4-cyclohexyl, lvijlerexmvran-uhfffaoysa
PubChem CID 90718
IUPAC Name 1-bromo-4-cyclohexylbenzene
SMILES C1CCC(CC1)C2=CC=C(C=C2)Br

Specifications

Density 1.287
Boiling Point 90°C to 94°C (0.01 mmHg)
Refractive Index 1.5595
Quantity 5 g
Beilstein 2209869
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 239.16
Percent Purity 98%
Chemical Name or Material 1-Bromo-4-cyclohexylbenzene
EINECSNumber 246-623-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only