missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-4-butoxybenzene, 97%, Thermo Scientific™
$98.86 - $98.86
Chemical Identifiers
| CAS | 39969-57-8 |
|---|---|
| Molecular Formula | C10H13BrO |
| Molecular Weight (g/mol) | 229.117 |
| MDL Number | MFCD00037100 |
| InChI Key | BOUVKHWPQNEXTO-UHFFFAOYSA-N |
| PubChem CID | 142395 |
| IUPAC Name | 1-bromo-4-butoxybenzene |
| SMILES | CCCCOC1=CC=C(C=C1)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
BTB09793EA
|
Thermo Scientific Chemicals
BTB09793EA |
10 g |
N/A
|
|
|||||
Chemical Identifiers
| 39969-57-8 | |
| 229.117 | |
| BOUVKHWPQNEXTO-UHFFFAOYSA-N | |
| 1-bromo-4-butoxybenzene |
| C10H13BrO | |
| MFCD00037100 | |
| 142395 | |
| CCCCOC1=CC=C(C=C1)Br |
Specifications
| 39969-57-8 | |
| 100.0 | |
| Amber glass bottle | |
| BrC6H4O(CH2)3CH3 | |
| 10 g | |
| BOUVKHWPQNEXTO-UHFFFAOYSA-N | |
| 1-bromo-4-butoxybenzene | |
| 142395 | |
| 97% |
| 96.5 | |
| 97% | |
| C10H13BrO | |
| MFCD00037100 | |
| p-bromophenyl butyl ether, benzene,1-bromo-4-butoxy, p-butoxybromobenzene, benzene, 1-bromo-4-butoxy, 4-bromo-1-butoxybenzene, 4-butoxybromobenzene, 4-n-butoxyphenyl bromide, 1-butoxy-4-bromobenzene, 4-bromophenyl butyl ether | |
| CCCCOC1=CC=C(C=C1)Br | |
| 229.117 | |
| 229.12 | |
| 1-Bromo-4-butoxybenzene |