accessibility menu, dialog, popup

UserName

CAS RN 39969-57-8

IUPAC Name: 1-bromo-4-butoxybenzene
Synonyms: 1-bromo-4-butoxybenzene
Molecular Weight (g/mol): 229.12
Molecular Formula: C10H13BrO
InChi Key: BOUVKHWPQNEXTO-UHFFFAOYSA-N
SMILES: CCCCOC1=CC=C(Br)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results