missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-methylbutane 96.0+%, TCI America™

Catalog No. p-7131449
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I050425G 25 g
I0504500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I050425G Supplier TCI America Supplier No. I050425G
Only null left

Chemical Identifiers

CAS 107-82-4
Molecular Formula C5H11Br
Molecular Weight (g/mol) 151.05
MDL Number MFCD00000253
InChI Key YXZFFTJAHVMMLF-UHFFFAOYSA-N
Synonym isoamyl bromide, isopentyl bromide, butane, 1-bromo-3-methyl, 3-methylbutyl bromide, 4-bromo-2-methylbutane, isobutylmethyl bromide, isopentylbromide, 1-bromo-3-methyl-butane, iso-pentyl bromide, unii-czm50594qa
PubChem CID 7891
IUPAC Name 1-bromo-3-methylbutane
SMILES CC(C)CCBr

Specifications

Color Yellow
Boiling Point 121°C
Quantity 25 g
UN Number 2341
Formula Weight 151.05
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-3-methylbutane
EINECSNumber (2)-0067
RTECSNumber EJ6230000
TSCA Yes