accessibility menu, dialog, popup

UserName

CAS RN 107-82-4

IUPAC Name: 1-bromo-3-methylbutane
Synonyms: 1-bromo-3-methylbutane
Molecular Weight (g/mol): 151.05
Molecular Formula: C5H11Br
InChi Key: YXZFFTJAHVMMLF-UHFFFAOYSA-N
SMILES: CC(C)CCBr
Boiling Point 
  • (6)
  • (2)
Percent Purity 
  • (2)
  • (4)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (6)
  • (6)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)