Learn More
1-Bromo-3-methylbutane, 99%
CAS: 107-82-4 | C5H11Br | 151.05 g/mol
$60.19 - $60.19
Chemical Identifiers
| CAS | 107-82-4 |
|---|---|
| Molecular Formula | C5H11Br |
| Molecular Weight (g/mol) | 151.05 |
| MDL Number | MFCD00000253 |
| InChI Key | YXZFFTJAHVMMLF-UHFFFAOYSA-N |
| Synonym | isoamyl bromide, isopentyl bromide, butane, 1-bromo-3-methyl, 3-methylbutyl bromide, 4-bromo-2-methylbutane, isobutylmethyl bromide, isopentylbromide, 1-bromo-3-methyl-butane, iso-pentyl bromide, unii-czm50594qa |
| PubChem CID | 7891 |
| IUPAC Name | 1-bromo-3-methylbutane |
| SMILES | CC(C)CCBr |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC403051000
|
Thermo Scientific Chemicals
403051000 |
100 g | Glass bottle |
Each for $60.19
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 107-82-4 | |
| 151.05 | |
| YXZFFTJAHVMMLF-UHFFFAOYSA-N | |
| 7891 | |
| CC(C)CCBr |
| C5H11Br | |
| MFCD00000253 | |
| isoamyl bromide, isopentyl bromide, butane, 1-bromo-3-methyl, 3-methylbutyl bromide, 4-bromo-2-methylbutane, isobutylmethyl bromide, isopentylbromide, 1-bromo-3-methyl-butane, iso-pentyl bromide, unii-czm50594qa | |
| 1-bromo-3-methylbutane |
Specifications
| 107-82-4 | |
| 98.5 | |
| -112°C | |
| 1.2000g/mL | |
| 32°C | |
| 98.5% min. (GC) | |
| C5H11Br | |
| (CH3)2CHCH2CH2Br | |
| 100 g | |
| 1.2 | |
| isoamyl bromide, isopentyl bromide, butane, 1-bromo-3-methyl, 3-methylbutyl bromide, 4-bromo-2-methylbutane, isobutylmethyl bromide, isopentylbromide, 1-bromo-3-methyl-butane, iso-pentyl bromide, unii-czm50594qa | |
| YXZFFTJAHVMMLF-UHFFFAOYSA-N | |
| 1-bromo-3-methylbutane | |
| 7891 | |
| 99% | |
| 1-Bromo-3-methylbutane, 99% |
| 0.1% max. (as HBr) | |
| 100.0 | |
| Colorless | |
| 120°C to 121°C | |
| Authentic | |
| Glass bottle | |
| 1.4399 to 1.4419 | |
| MFCD00000253 | |
| 01, 136 | |
| 15, 5159 | |
| Solubility in water: 0.2g/L in water (16°C). Other solubilities: miscible with alcohol,ether,benzene and,carbon tetrachloride | |
| CC(C)CCBr | |
| 151.05 | |
| 151.05 | |
| Liquid |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN: Wash with plenty of soap a
GHS Signal Word: Warning
EINECSNumber : 203-522-9
RTECSNumber : EJ6230000
TSCA : TSCA
RUO – Research Use Only