missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-methylbutane, 99%

Catalog No. AC403051000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC403051000 100 g Glass bottle
AC403055000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC403051000 Supplier Thermo Scientific Chemicals Supplier No. 403051000

CAS: 107-82-4 | C5H11Br | 151.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 107-82-4
Molecular Formula C5H11Br
Molecular Weight (g/mol) 151.05
MDL Number MFCD00000253
InChI Key YXZFFTJAHVMMLF-UHFFFAOYSA-N
Synonym isoamyl bromide, isopentyl bromide, butane, 1-bromo-3-methyl, 3-methylbutyl bromide, 4-bromo-2-methylbutane, isobutylmethyl bromide, isopentylbromide, 1-bromo-3-methyl-butane, iso-pentyl bromide, unii-czm50594qa
PubChem CID 7891
IUPAC Name 1-bromo-3-methylbutane
SMILES CC(C)CCBr

Specifications

Acidity 0.1% max. (as HBr)
CAS Min % 98.5
CAS Max % 100.0
Melting Point -112°C
Color Colorless
Density 1.2000g/mL
Boiling Point 120°C to 121°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4399 to 1.4419
Linear Formula (CH3)2CHCH2CH2Br
Quantity 100 g
Beilstein 01, 136
Specific Gravity 1.2
Merck Index 15, 5159
Solubility Information Solubility in water: 0.2g/L in water (16°C). Other solubilities: miscible with alcohol,ether,benzene and,carbon tetrachloride
Formula Weight 151.05
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Bromo-3-methylbutane, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN: Wash with plenty of soap a
EINECSNumber 203-522-9
RTECSNumber EJ6230000
TSCA TSCA

RUO – Research Use Only