missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-methylbutane, 98%

CAS: 107-82-4 | C5H11Br | 151.05 g/mol

Supplier:  Thermo Scientific Chemicals A1449230

 View more versions of this product

Catalog No. AAA1449230

Only null left
Explore available promotions

Description

Description

1-Bromo-3-methylbutane was used in the synthesis of 1-(3-methylbutyl)pyrrole and in the production of pentyl peroxy radical by direct photolysis production method. It was also used as extraction solvent for determination of polycyclic aromatic hydrocarbons in water samples using liquid-liquid microextraction combined with gas chromatography-mass spectrometry. It is also used as an alkyl halide and organic synthesis reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-3-methylbutane was used in the synthesis of 1-(3-methylbutyl)pyrrole and in the production of pentyl peroxy radical by direct photolysis production method. It was also used as extraction solvent for determination of polycyclic aromatic hydrocarbons in water samples using liquid-liquid microextraction combined with gas chromatography-mass spectrometry. It is also used as an alkyl halide and organic synthesis reagent.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

1-Bromo-3-methylbutane
-112°C
120°C to 121°C
C5H11Br
MFCD00000253
UN2341
14,5114
Not miscible or difficult to mix in water.
CC(C)CCBr
151.05
151.05
107-82-4
1.208
22°C (71°F)
1.4409
250 g
773652
isoamyl bromide, isopentyl bromide, butane, 1-bromo-3-methyl, 3-methylbutyl bromide, 4-bromo-2-methylbutane, isobutylmethyl bromide, isopentylbromide, 1-bromo-3-methyl-butane, iso-pentyl bromide, unii-czm50594qa
YXZFFTJAHVMMLF-UHFFFAOYSA-N
1-bromo-3-methylbutane
7891
98%

Chemical Identifiers

107-82-4
151.05
YXZFFTJAHVMMLF-UHFFFAOYSA-N
7891
CC(C)CCBr
C5H11Br
MFCD00000253
isoamyl bromide, isopentyl bromide, butane, 1-bromo-3-methyl, 3-methylbutyl bromide, 4-bromo-2-methylbutane, isobutylmethyl bromide, isopentylbromide, 1-bromo-3-methyl-butane, iso-pentyl bromide, unii-czm50594qa
1-bromo-3-methylbutane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: 1-BROMO-3-METHYLBUTANE

EINECSNumber : 203-522-9

RTECSNumber : EJ6230000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only