Learn More
1-Bromo-3-methylbutane, 98%
CAS: 107-82-4 | C5H11Br | 151.05 g/mol
Supplier: Thermo Scientific Chemicals A144920I
Description
1-Bromo-3-methylbutane was used in the synthesis of 1-(3-methylbutyl)pyrrole and in the production of pentyl peroxy radical by direct photolysis production method. It was also used as extraction solvent for determination of polycyclic aromatic hydrocarbons in water samples using liquid-liquid microextraction combined with gas chromatography-mass spectrometry. It is also used as an alkyl halide and organic synthesis reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromo-3-methylbutane was used in the synthesis of 1-(3-methylbutyl)pyrrole and in the production of pentyl peroxy radical by direct photolysis production method. It was also used as extraction solvent for determination of polycyclic aromatic hydrocarbons in water samples using liquid-liquid microextraction combined with gas chromatography-mass spectrometry. It is also used as an alkyl halide and organic synthesis reagent.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
| 1-Bromo-3-methylbutane | |
| -112°C | |
| 120°C to 121°C | |
| C5H11Br | |
| MFCD00000253 | |
| UN2341 | |
| 14,5114 | |
| Not miscible or difficult to mix in water. | |
| CC(C)CCBr | |
| 151.05 | |
| 151.05 |
| 107-82-4 | |
| 1.208 | |
| 22°C (71°F) | |
| 1.4409 | |
| 5000 g | |
| 773652 | |
| isoamyl bromide, isopentyl bromide, butane, 1-bromo-3-methyl, 3-methylbutyl bromide, 4-bromo-2-methylbutane, isobutylmethyl bromide, isopentylbromide, 1-bromo-3-methyl-butane, iso-pentyl bromide, unii-czm50594qa | |
| YXZFFTJAHVMMLF-UHFFFAOYSA-N | |
| 1-bromo-3-methylbutane | |
| 7891 | |
| 98% |
Chemical Identifiers
| 107-82-4 | |
| 151.05 | |
| YXZFFTJAHVMMLF-UHFFFAOYSA-N | |
| 7891 | |
| CC(C)CCBr |
| C5H11Br | |
| MFCD00000253 | |
| isoamyl bromide, isopentyl bromide, butane, 1-bromo-3-methyl, 3-methylbutyl bromide, 4-bromo-2-methylbutane, isobutylmethyl bromide, isopentylbromide, 1-bromo-3-methyl-butane, iso-pentyl bromide, unii-czm50594qa | |
| 1-bromo-3-methylbutane |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: 1-BROMO-3-METHYLBUTANE
EINECSNumber : 203-522-9
RTECSNumber : EJ6230000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only