missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-isopropoxybenzene 98.0+%, TCI America™

Catalog No. p-7119546
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21985G 5 g
B219825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21985G Supplier TCI America Supplier No. B21985G

Chemical Identifiers

CAS 131738-73-3
Molecular Formula C9H11BrO
Molecular Weight (g/mol) 215.09
MDL Number MFCD00070756
InChI Key JYWJZIQSPRFCDB-UHFFFAOYSA-N
Synonym 1-bromo-3-isopropoxybenzene, 3-bromophenyl isopropyl ether, 1-bromo-3-propan-2-yloxy benzene, 3-bromo-isopropoxybenzene, 3-bromoisoproproxylbenzene, 2-3-bromophenoxy propane, 3-bromoisopropoxybenzene, 3-bromo phenyl isopropyl ether, benzene, 1-bromo-3-1-methylethoxy, pubchem14867
PubChem CID 3260801
IUPAC Name 1-bromo-3-(propan-2-yloxy)benzene
SMILES CC(C)OC1=CC(Br)=CC=C1

Specifications

Color Colorless
Quantity 5 g
Formula Weight 215.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-3-isopropoxybenzene
EINECSNumber (3)-0845
TSCA No